Learn More
2,3-Dihydro-3,5-dihydroxy-6-methyl-4-pyrone, TRC
Product Code. 30357110
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357110 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30357110 Supplier Toronto Research Chemicals Supplier No. TRCD28564010MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,3-Dihydro-3,5-dihydroxy-6-methyl-4-pyrone
CAS 28564-83-2
Molecular Formula C6H8O4
Quantity 10 mg
Synonym 2,3-Dihydro-3,5-dihydroxy-6-methyl-4-pyranone, 3-Hydroxy-2,3-dihydromaltol, 5-Hydroxy-5,6-dihydromaltol, 6-Methyl-3,5-dihydroxy-2,3-dihydro-4H-pyran-4-one, DDMP, Pyranone
InChI Formula InChI=1S/C6H8O4/c1-3-5(8)6(9)4(7)2-10-3/h4,7-8H,2H2,1H3
SMILES CC1=C(O)C(=O)C(O)CO1
IUPAC Name 3,5-dihydroxy-6-methyl-2,3-dihydropyran-4-one
Molecular Weight (g/mol) 144.13
Formula Weight 144.0423