missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydro-2-methyl-1H-indol-1-amine Hydrochloride, TRC

Product Code. 30373394
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373394 1 g
30382187 50 g
30371841 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30373394 Supplier Toronto Research Chemicals Supplier No. TRCD4496451G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102789-79-7
Molecular Formula C9 H12 N2 . Cl H
Molecular Weight (g/mol) 184.67
Synonym 1H-Indol-1-amine, 2,3-dihydro-2-methyl-, hydrochloride (9CI), 1-Amino-2-methylindoline Hydrochloride
IUPAC Name 2-methyl-2,3-dihydroindol-1-amine;hydrochloride
SMILES Cl.CC1Cc2ccccc2N1N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H12N2.ClH/c1-7-6-8-4-2-3-5-9(8)11(7)10;/h2-5,7H,6,10H2,1H3;1H
Formula Weight 184.0767
Chemical Name or Material 2-Methylindolin-1-amine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.