missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzonitrile, 98%, Thermo Scientific™

Product Code. 10492641 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10492641

Brand: Thermo Scientific Acros 303660010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21524-39-0
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.105
MDL Number MFCD00009976
InChI Key GKPHNZYMLJPYJJ-UHFFFAOYSA-N
Synonym benzonitrile, 2,3-difluoro, 2,3-difluorobenzenecarbonitrile, ncr bf cf, difluorobenzonitrile, pubchem1564, acmc-209flg, 2,3-difluorobenzonitril, 2,3-difluoro-benzonitrile, 2.3-difluoro benzonitrile, ksc493a8b
PubChem CID 88935
IUPAC Name 2,3-difluorobenzonitrile
SMILES C1=CC(=C(C(=C1)F)F)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.25g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula FC6H3CN
Refractive Index 1.4872 to 1.4892 (20°C, 589nm)
Quantity 1 g
UN Number 2810
Formula Weight 139.11
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2, 3-Difluorobenzonitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.