missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorobenzaldehyde, 98%, Thermo Scientific™

Product Code. 11490591
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490591 1 g 1g
11400601 5 g 5g
11454110 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490591 Supplier Thermo Scientific Alfa Aesar Supplier No. B20093.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2646-91-5
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.105
MDL Number MFCD00010292
InChI Key WDBAXYQUOZDFOJ-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3-difluoro, 2,3-difluoro benzaldehyde, 2,3-difluoro-benzaldehyde, difluoro-benzaldehyde, pubchem1436, 2,3 difluorobenzaldehyde, acmc-1cr6r, 2,3-difluorobenzaldehyde?, 2,3-difluorobenzaldehyde, ksc493k8r
PubChem CID 137664
IUPAC Name 2,3-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)F)F)C=O

Specifications

Density 1.301
Boiling Point 69°C to 70°C (15 mmHg)
Flash Point 58°C (136°F)
Refractive Index 1.5
Quantity 1 g
UN Number UN1989
Beilstein 2961554
Sensitivity Air Sensitive
Formula Weight 142.11
Percent Purity 98%
Chemical Name or Material 2,3-Difluorobenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.