Learn More
2,3-Diethyl-5-methylpyrazine, 98+%, Thermo Scientific™
Product Code. 11427637
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11427637
Brand: Thermo Scientific Alfa Aesar A15686.22
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18138-04-0
Molecular Formula C9H14N2
Molecular Weight (g/mol) 150.23
MDL Number MFCD00009635
InChI Key PSINWXIDJYEXLO-UHFFFAOYSA-N
Synonym pyrazine, 2,3-diethyl-5-methyl, 2-methyl-5,6-diethylpyrazine, 2,3-diethyl-6-methylpyrazine, 5-methyl-2,3-diethylpyrazine, 2,3-diethyl-5-methyl-pyrazine, fema no. 3336, hazelnut pyrazine, pubchem8621, acmc-1ca9d, ksc491g2h
PubChem CID 28905
IUPAC Name 2,3-diethyl-5-methylpyrazine
SMILES CCC1=NC=C(C)N=C1CC
Specifications
Density 0.949
Boiling Point 190°C
Flash Point 80°C (176°F)
Refractive Index 1.498
Quantity 100 g
Formula Weight 150.23
Percent Purity ≥98%
Chemical Name or Material 2,3-Diethyl-5-methylpyrazine