Learn More
2,3-Dichlorobenzonitrile, 98%
Product Code. p-7022173
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11462557
Brand: Thermo Scientific Alfa Aesar A14809.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 6574-97-6 | C7H3Cl2N | 172.008 g/mol
2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Dichlorobenzonitrile, is used as a reactant involved in nucleophilic aromatic fluorination, reactions with magnesium amides for synthesis of carboxamides, Suzuki-Miyaura coupling reactions. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
Solubility
It is miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6574-97-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00016372
InChI Key OHDYZVVLNPXKDX-UHFFFAOYSA-N
Synonym benzonitrile, dichloro, 2,3-dichlorobenzenecarbonitrile, benzonitrile, 2,3-dichloro, dichlorobenzonitrile, pubchem12516, 2,3-dichloro benzonitrile, 2,3-dichlorophenyl cyanide, 2,3-di-chloro-benzonitrile, acmc-1b38y, ksc357s4b
PubChem CID 736567
IUPAC Name 2,3-dichlorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)Cl)C#N
Specifications
Melting Point 60°C to 65°C
Boiling Point 114°C (6 mmHg)
Quantity 5 g
UN Number UN3439
Beilstein 2438753
Solubility Information It is miscible with water.
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzonitrile