Learn More
2,3-Dichloro-5-fluoropyridine, TRC
Product Code. 30370133
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370133 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30370133 Supplier Toronto Research Chemicals Supplier No. TRCD434395250MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,3-Dichloro-5-fluoropyridine
CAS 185985-40-4
Molecular Formula C5H2Cl2FN
Quantity 250 mg
Synonym 2,3-Dichloro-5-fluoro-pyridine
InChI Formula InChI=1S/C5H2Cl2FN/c6-4-1-3(8)2-9-5(4)7/h1-2H
SMILES FC1=CN=C(Cl)C(Cl)=C1
IUPAC Name 2,3-dichloro-5-fluoropyridine
Molecular Weight (g/mol) 165.98
Formula Weight 164.95