Learn More
2,3-Dichloro-5-fluoropyridine, TRC
Product Code. 30374375
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30374375 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30374375 Supplier Toronto Research Chemicals Supplier No. TRCD4343952.5G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,3-Dichloro-5-fluoropyridine
CAS 185985-40-4
Molecular Formula C5H2Cl2FN
Quantity 2.5 g
Synonym 2,3-Dichloro-5-fluoro-pyridine
InChI Formula InChI=1S/C5H2Cl2FN/c6-4-1-3(8)2-9-5(4)7/h1-2H
SMILES FC1=CN=C(Cl)C(Cl)=C1
IUPAC Name 2,3-dichloro-5-fluoropyridine
Molecular Weight (g/mol) 165.98
Formula Weight 164.95