Learn More
2-[(3-Carboxyphenyl)amino]-5-nitrobenzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-[(3-Carboxyphenyl)amino]-5-nitrobenzoic Acid |
| CAS | 2770903-44-9 |
| Molecular Formula | C14H10N2O7 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C14H10N2O7/c17-12-4-1-7(5-10(12)14(20)21)15-11-3-2-8(16(22)23)6-9(11)13(18)19/h1-6,15,17H,(H,18,19)(H,20,21) |
| SMILES | OC1=C(C(O)=O)C=C(NC(C(C(O)=O)=C2)=CC=C2[N+]([O-])=O)C=C1 |
| IUPAC Name | 2-((3-carboxy-4-hydroxyphenyl)amino)-5-nitrobenzoic acid |
| Molecular Weight (g/mol) | 318.24 |
| Formula Weight | 318.05 |