Learn More
2,3,5-Trimethylphenol-d11, CDN
Specifications
Specifications
| Chemical Name or Material | 2,3,5-Trimethylphenol-d11 |
| CAS | 362049-46-5 |
| Molecular Formula | C9 D11 H O |
| Quantity | 0.5 g |
| Synonym | Phen-2,4-d2-ol, 3,5,6-tri(methyl-d3)- (9 CI), 2,3,5-Trimethylphenol-d11, 2,3,5-Trimethylphenol-D11, 2,3,5-Tri(methyl-d3)phenol-4,6-d2, 3,5,6-Tri(methyl-d3)phenol-2,4-d2 |
| InChI Formula | InChI=1 S/C9H12O/c1-6-4-7(2)8(3)9(10)5-6/h4-5,10 H,1-3H3/i1D3,2D3,3D3,4 D,5 D |
| SMILES | [2 H]c1c(O)c(c(c([2 H])c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,4-dideuterio-3,5,6-tris(trideuteriomethyl)phenol |
| Molecular Weight (g/mol) | 147.2588 |
| Formula Weight | 147.1579 g/mol |
| Show More |