missing translation for 'onlineSavingsMsg'
Learn More

2’,3’,4’-Tri-O-benzoyl Propofol-d17 Beta-D-Glucuronide Methyl Ester, TRC

Product Code. 30455983
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455983 10 mg 10mg
30449718 2.5 mg 2.50mg
30451156 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30455983 Supplier Toronto Research Chemicals Supplier No. TRCT76800710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2’,3’,4’-Tri-O-benzoyl Propofol-d17 beta-D-Glucuronide Methyl Ester
Molecular Formula C40H23D17O10
Quantity 10 mg
InChI Formula InChI=1S/C40H40O10/c1-24(2)29-22-15-23-30(25(3)4)31(29)49-40-35(48-38(43)28-20-13-8-14-21-28)33(47-37(42)27-18-11-7-12-19-27)32(34(50-40)39(44)45-5)46-36(41)26-16-9-6-10-17-26/h6-25,32-35,40H,1-5H3/t32?,33?,34?,35?,40-/m1/s1/i1D3,2D3,3D3,4D3,15D,22D,23D,24D,25D
SMILES [2H]C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C1=C([2H])C([2H])=C([2H])C(C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=C1O[C@@H]2O[C@H](C(OC)=O)[C@@H](OC(C3=CC=CC=C3)=O)[C@H](OC(C4=CC=CC=C4)=O)[C@H]2OC(C5=CC=CC=C5)=O
IUPAC Name (2S,3R,4S,5S,6S)-2-(2,6-bis(propan-2-yl-d7)phenoxy-3,4,5-d3)-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl tribenzoate
Molecular Weight (g/mol) 697.84
Formula Weight 697.37
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.