Learn More
2-(3,4-Dimethoxybenzoyl)pyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(3,4-Dimethoxybenzoyl)pyridine |
| CAS | 27693-42-1 |
| Molecular Formula | C14H13NO3 |
| Quantity | 500 mg |
| Synonym | (3,4-Dimethoxyphenyl)-2-pyridinylmethanone, 3,4-Dimethoxyphenyl 2-Pyridyl Ketone |
| InChI Formula | InChI=1S/C14H13NO3/c1-17-12-7-6-10(9-13(12)18-2)14(16)11-5-3-4-8-15-11/h3-9H,1-2H3 |
| SMILES | O=C(C1=NC=CC=C1)C2=CC=C(OC)C(OC)=C2 |
| IUPAC Name | (3,4-dimethoxyphenyl)(pyridin-2-yl)methanone |
| Molecular Weight (g/mol) | 243.26 |
| Formula Weight | 243.09 |