Learn More
2,3,4,5-Tetrahydro-1H-1,5-methano-3-benzazepine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3,4,5-Tetrahydro-1H-1,5-methano-3-benzazepine hydrochloride |
| CAS | 230615-52-8 |
| Molecular Formula | C11 H13 N . Cl H |
| Quantity | 500 mg |
| Synonym | 1,5-Methano-1H-3-benzazepine','2,3,4,5-tetrahydro-','hydrochloride (1:1), 1,5-Methano-1H-3-benzazepine','2,3,4,5-tetrahydro-','hydrochloride, 2,3,4,5-Tetrahydro-1,5-methano-1H-3-benzazepine hydrochloride |
| InChI Formula | InChI=1S/C11H13N.ClH/c1-2-4-11-9-5-8(6-12-7-9)10(11)3-1;/h1-4,8-9,12H,5-7H2;1H |
| SMILES | Cl.C1NCC2CC1c3ccccc23 |
| IUPAC Name | (1S,8R)-10-azatricyclo[6.3.1.02,7]dodeca-2,4,6-triene;hydrochloride |
| Molecular Weight (g/mol) | 195.69 |
| Formula Weight | 195.0815 |