Learn More
2,3,4,5,6-Pentabromotoluene-13C6, TRC
Specifications
Specifications
| Chemical Name or Material | 2,3,4,5,6-Pentabromotoluene-13C6 |
| CAS | 2724878-50-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C13C6H3Br5 |
| Quantity | 10 mg |
| Synonym | Pentabromotoluene-13C6 |
| InChI Formula | InChI=1S/C7H3Br5/c1-2-3(8)5(10)7(12)6(11)4(2)9/h1H3/i2+1,3+1,4+1,5+1,6+1,7+1 |
| SMILES | Br[13C]1=[13C](Br)[13C](Br)=[13C](C)[13C](Br)=[13C]1Br |
| IUPAC Name | 1,2,3,4,5-pentabromo-6-methylbenzene-1,2,3,4,5,6-13C6 |
| Molecular Weight (g/mol) | 492.57 |
| Show More |