Learn More
2,2-Dimethyl Metolazone, TRC
Specifications
Specifications
| CAS | 28638-47-3 |
| Molecular Formula | C16H16ClN3O3S |
| Quantity | 500 mg |
| Synonym | 7-Chloro-1,2,3,4-tetrahydro-2,2-dimethyl-4-oxo-3-phenyl-6-quinazolinesulfonamide |
| InChI Formula | InChI=1S/C16H16ClN3O3S/c1-16(2)19-13-9-12(17)14(24(18,22)23)8-11(13)15(21)20(16)10-6-4-3-5-7-10/h3-9,19H,1-2H3,(H2,18,22,23) |
| SMILES | CC1(C)Nc2cc(Cl)c(cc2C(=O)N1c3ccccc3)S(=O)(=O)N |
| IUPAC Name | 7-chloro-2,2-dimethyl-4-oxo-3-phenyl-1H-quinazoline-6-sulfonamide |
| Molecular Weight (g/mol) | 365.83 |
| Formula Weight | 365.0601 |
| Chemical Name or Material | 2,2-Dimethyl Metolazone |