missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-6-nitro-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific Chemicals

Product Code. p-7041139
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476266 250 mg 250mg
15466266 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476266 Supplier Thermo Scientific Alfa Aesar Supplier No. H61097.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85160-84-5
Molecular Formula C10H10N2O4
Molecular Weight (g/mol) 222.20
MDL Number MFCD03427729
InChI Key YKXZRZGZJZYBBH-UHFFFAOYSA-N
Synonym 2,2-dimethyl-6-nitro-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl-6-nitro, 2,2-dimethyl-6-nitro-2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl-6-nitro-4h-benzo 1,4 oxazin-3-one, 2,2-dimethyl-6-nitro-2,3-dihydro-4h-1,4-benzooxazine-3-one, 3,4-dihydro-2,2-dimethyl-6-nitro-3-oxo-2h-1,4-benzoxazine, 2,2-dimethyl-6-nitro-2h-benzo b 1.4 oxazine-3 4h-one, 2,2-dimethyl-6-nitro-3,4-dihydro-2h-benzo 1,4 oxazine-one
PubChem CID 10656631
IUPAC Name 2,2-dimethyl-6-nitro-4H-1,4-benzoxazin-3-one
SMILES CC1(C)OC2=C(NC1=O)C=C(C=C2)[N+]([O-])=O

Specifications

Quantity 250 mg
Formula Weight 222.2
Percent Purity 97%
Chemical Name or Material 2,2-Dimethyl-6-nitro-2H-1,4-benzoxazin-3(4H)-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.