Learn More
2,2-dimethyl-4-oxo-4-(propan-2-yloxy)butanoic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2-dimethyl-4-oxo-4-(propan-2-yloxy)butanoic acid |
| CAS | 1803601-41-3 |
| Molecular Formula | C9H16O4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C9H16O4/c1-6(2)13-7(10)5-9(3,4)8(11)12/h6H,5H2,1-4H3,(H,11,12) |
| SMILES | CC(C)OC(=O)CC(C)(C)C(=O)O |
| IUPAC Name | 2,2-dimethyl-4-oxo-4-propan-2-yloxybutanoic acid |
| Molecular Weight (g/mol) | 188.22 |
| Formula Weight | 188.1049 |