Learn More
2,2'-Dimethoxy-1,1'-biphenyl, TRC
Product Code. 30373520
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373520 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30373520 Supplier Toronto Research Chemicals Supplier No. TRCD46048050MG
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,2'-Dimethoxy-1,1'-biphenyl
CAS 4877-93-4
Molecular Formula C14 H14 O2
Quantity 50 mg
InChI Formula InChI=1S/C14H14O2/c1-15-13-9-5-3-7-11(13)12-8-4-6-10-14(12)16-2/h3-10H,1-2H3
SMILES COc1ccccc1c2ccccc2OC
IUPAC Name 1-methoxy-2-(2-methoxyphenyl)benzene
Molecular Weight (g/mol) 214.26
Formula Weight 214.0994
Product Title
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.