missing translation for 'onlineSavingsMsg'
Learn More

(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%

Product Code. 15408295
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408295 1 g 1g
15418295 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408295 Supplier Thermo Scientific Alfa Aesar Supplier No. H33055.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 128073-33-6 | C5H6F2O2 | 136.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128073-33-6
Molecular Formula C5H6F2O2
Molecular Weight (g/mol) 136.098
MDL Number MFCD07777163
InChI Key HLFLYOQLHYYNLT-UHFFFAOYSA-N
Synonym 2,2-difluoro-1-methylcyclopropanecarboxylic acid, -2,2-difluoro-1-methylcyclopropanecarboxylic acid, +/--2,2-difluoro-1-methylcyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane carboxylic acid, 2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid, 2,2-difluoro-1-methyl-cyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane-carboxylic acid, -2,2-difluoro-1-methylcyclopropane-carboxylicacid, 2,2-difluoro-1-methyl cyclopropane carboxylic acid, +/--2,2-difluoro-1-methylcyclopropane-carboxylicacid
PubChem CID 14695229
IUPAC Name 2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
SMILES CC1(CC1(F)F)C(=O)O

Specifications

Melting Point 62°C to 64°C
Quantity 1 g
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material (±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.