missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid, 97%

Product Code. p-7027302
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11406836

Brand: Thermo Scientific Alfa Aesar B23955.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126120-85-2 | C8H4F2O4 | 202.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126120-85-2
Molecular Formula C8H4F2O4
Molecular Weight (g/mol) 202.113
MDL Number MFCD01631473
InChI Key ZGAQVJDFFVTWJK-UHFFFAOYSA-N
Synonym 2,2-difluorobenzo d 1,3 dioxole-4-carboxylic acid, 2,2-difluoro-2h-1,3-benzodioxole-4-carboxylic acid, 2,2-difluoro-1,3-benzodioxole-4-carboxylicacid, 2,2-difluoro-benzo 1,3 dioxole-4-carboxylic acid, 2,2-difluorobenzo d 1,3-dioxolene-4-carboxylic acid, acmc-1c3im, 1,3-benzodioxole-4-carboxylicacid, 2,2-difluoro, 2,2-bis fluoranyl-1,3-benzodioxole-4-carboxylic acid, 2,2-difluoro-1,3-benzodioxole-4-carboxylic acid
PubChem CID 2774067
IUPAC Name 2,2-difluoro-1,3-benzodioxole-4-carboxylic acid
SMILES C1=CC(=C2C(=C1)OC(O2)(F)F)C(=O)O

Specifications

Melting Point 202°C to 205°C
Quantity 1 g
Beilstein 7292629
Formula Weight 202.12
Percent Purity 97%
Chemical Name or Material 2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.