Learn More
2,2-Diethylhexanoic Acid, TRC
Product Code. 30384328
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384328 5 g 5g
30380330 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30384328 Supplier Toronto Research Chemicals Supplier No. TRCD4444905G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4528-37-4
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.26
Synonym α,α-Diethyl-Caproic Acid, 2,2-Diethylhexanoic Acid, NSC 467, α,α-Diethylcaproic Acid
IUPAC Name 2,2-diethylhexanoic acid
SMILES O=C(O)C(CC)(CC)CCCC
Specifications
Quantity 5 g
InChI Formula InChI=1S/C10H20O2/c1-4-7-8-10(5-2,6-3)9(11)12/h4-8H2,1-3H3,(H,11,12)
Formula Weight 172.1463
Chemical Name or Material 2,2-Diethylhexanoic Acid