Learn More
2,2'-Dichloro Diphenyl Disulfide, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2'-Dichloro Diphenyl Disulfide |
| CAS | 31121-19-4 |
| Molecular Formula | C12H8Cl2S2 |
| Quantity | 1 g |
| Synonym | Bis(2-chlorophenyl) Disulfide, Bis(o-chlorophenyl) Disulfide, NSC 32024, 1,2-Bis(2-chlorophenyl)disulfane |
| InChI Formula | InChI=1S/C12H8Cl2S2/c13-9-5-1-3-7-11(9)15-16-12-8-4-2-6-10(12)14/h1-8H |
| SMILES | Clc1ccccc1SSc2ccccc2Cl |
| Molecular Weight (g/mol) | 287.23 |
| Formula Weight | 285.9444 |