missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chlorophenoxy)ethanol, 98%, Thermo Scientific™

Product Code. 11326916
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326916 5 g 5g
11336916 25 g 25g
11346916 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326916 Supplier Thermo Scientific Alfa Aesar Supplier No. L04368.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15480-00-9
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.608
MDL Number MFCD00014034
InChI Key FDQGMCQSIVZGHW-UHFFFAOYSA-N
PubChem CID 84933
IUPAC Name 2-(2-chlorophenoxy)ethanol
SMILES C1=CC=C(C(=C1)OCCO)Cl

Specifications

Density 1.257
Quantity 5 g
Refractive Index 1.553
UN Number UN2810
Beilstein 1945576
Synonym 2-2-chlorophenoxy ethanol, ethanol, 2-2-chlorophenoxy, 2-2'-chlorophenoxy ethanol, 2-2-chlorophenoxy ethan-1-ol, ethyleneglycolmono2chlorophenylether, 2-chlorophenoxy ethanol, acmc-20aor0, ethanol,2-2-chlorophenoxy, 2-2-hydroxyethoxy-chlorobenzene
Boiling Point 114°C to 116°C (1 mmHg)
Formula Weight 172.61
Percent Purity 98%
Chemical Name or Material 2-(2-Chlorophenoxy)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.