Learn More
2-(2-Bromo-acetylamino)-5-chloro-benzophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(2-Bromo-acetylamino)-5-chloro-benzophenone |
| CAS | 32580-26-0 |
| Molecular Formula | C15 H11 Br Cl N O2 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C15H11BrClNO2/c16-9-14(19)18-13-7-6-11(17)8-12(13)15(20)10-4-2-1-3-5-10/h1-8H,9H2,(H,18,19) |
| SMILES | Clc1ccc(NC(=O)CBr)c(c1)C(=O)c2ccccc2 |
| IUPAC Name | N-(2-benzoyl-4-chlorophenyl)-2-bromoacetamide |
| Molecular Weight (g/mol) | 352.61 |
| Formula Weight | 350.9662 |