missing translation for 'onlineSavingsMsg'
Learn More

2,2,4,6,6-Pentamethylheptane, TRC

Product Code. 30011098
Change view
Click to view available options
Quantity:
1 mL
2.5 mL
5 mL
Unit Size:
1mL
2.5mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011098 5 mL 5mL
30136725 2.5 mL 2.5mL
30004141 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 30011098 Supplier Toronto Research Chemicals Supplier No. TRCP2272255ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13475-82-6
Formula Weight 170.2035
InChI Formula InChI=1S/C12H26/c1-10(8-11(2,3)4)9-12(5,6)7/h10H,8-9H2,1-7H3
IUPAC Name 2,2,4,6,6-pentamethylheptane
Molecular Formula C12 H26
Molecular Weight (g/mol) 170.33
SMILES CC(CC(C)(C)C)CC(C)(C)C
Synonym Sodium saccharin, Sodium 2-benzimidazole-1,1-dioxide
Chemical Name or Material 2,2,4,6,6-Pentamethylheptane

Specifications

Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.