Learn More
2,2,2-Trichloroethyl chloroformate, 97%
Product Code. 11387026
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387026 100 g 100g
11377026 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11387026 Supplier Thermo Scientific Alfa Aesar Supplier No. L06875.22
 Restricted Item
In Stock
Add to basket
CAS: 17341-93-4 | C3H2Cl4O2 | 211.847 g/mol
2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.
Solubility
Soluble in Ether, Benzene, Chloroform, Ethanol. Insoluble in water.
Notes
Moisture Sensitive. Desiccate at 4°C. Store under inert gas. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17341-93-4
Molecular Formula C3H2Cl4O2
Molecular Weight (g/mol) 211.847
MDL Number MFCD00000810
InChI Key LJCZNYWLQZZIOS-UHFFFAOYSA-N
Synonym 2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc
PubChem CID 87063
IUPAC Name 2,2,2-trichloroethyl carbonochloridate
SMILES C(C(Cl)(Cl)Cl)OC(=O)Cl
Specifications
Melting Point 0°C to 2°C
Density 1.539
Boiling Point 171°C to 172°C
Flash Point >100°C (212°F)
Refractive Index 1.47
Quantity 100 g
UN Number UN3277
Beilstein 970619
Sensitivity Moisture sensitive
Solubility Information Soluble in Ether,Benzene,Chloroform,Ethanol. Insoluble in water.
Formula Weight 211.86
Percent Purity 97%
Chemical Name or Material 2,2,2-Trichloroethyl chloroformate