Learn More
2-[2-(2’,6’-Dichloro-4’-hydroxphenylamino)phenyl]-N,N-dimethylacetamide, TRC
Product Code. 30375163
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375163 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30375163 Supplier Toronto Research Chemicals Supplier No. TRCD4346205MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-[2-(2',6'-Dichloro-4'-hydroxphenylamino)phenyl]-N,N-dimethylacetamide
CAS 698357-45-8
Molecular Formula C16 H16 Cl2 N2 O2
Quantity 5 mg
InChI Formula InChI=1S/C16H16Cl2N2O2/c1-20(2)15(22)7-10-5-3-4-6-14(10)19-16-12(17)8-11(21)9-13(16)18/h3-6,8-9,19,21H,7H2,1-2H3
SMILES CN(C)C(=O)Cc1ccccc1Nc2c(Cl)cc(O)cc2Cl
IUPAC Name 2-[2-(2,6-dichloro-4-hydroxyanilino)phenyl]-N,N-dimethylacetamide
Molecular Weight (g/mol) 339.22
Formula Weight 338.0589