Learn More
2’-[(1E)-2-[4-[[(2-Carboxyethyl)amino]carbonyl]phenyl]diazenyl] Balsalazide Trimethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 2'-((1E)-2-(4-(((2-Carboxyethyl)amino)carbonyl)phenyl)diazenyl) Balsalazide Trimethyl Ester |
| CAS | 2734467-53-7 |
| Molecular Formula | C30H30N6O9 |
| Quantity | 1 mg |
| Synonym | 2’-((1E)-2-(4-(((2-Carboxyethyl)amino)carbonyl)phenyl)diazenyl) Balsalazide Trimethyl Ester |
| InChI Formula | InChI=1S/C30H30N6O9/c1-43-26(38)12-14-31-28(40)18-4-7-20(8-5-18)33-36-24-16-19(29(41)32-15-13-27(39)44-2)6-10-23(24)35-34-21-9-11-25(37)22(17-21)30(42)45-3/h4-11,16-17,37H,12-15H2,1-3H3,(H,31,40)(H,32,41)/b35-34+,36-33+ |
| SMILES | OC1=C(C(OC)=O)C=C(/N=N/C2=C(/N=N/C3=CC=C(C(NCCC(OC)=O)=O)C=C3)C=C(C(NCCC(OC)=O)=O)C=C2)C=C1 |
| IUPAC Name | methyl 2-hydroxy-5-((E)-(4-((3-methoxy-3-oxopropyl)carbamoyl)-2-((E)-(4-((3-methoxy-3-oxopropyl)carbamoyl)phenyl)diazenyl)phenyl)diazenyl)benzoate |
| Molecular Weight (g/mol) | 618.59 |
| Formula Weight | 618.21 |