Learn More
2-(1-Nitroso-4-piperidinyl)-1H-benzimidazole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(1-Nitroso-4-piperidinyl)-1H-benzimidazole |
| CAS | 2731008-04-9 |
| Molecular Formula | C12 H14 N4 O |
| Quantity | 10 mg |
| Synonym | 2-(1-Nitroso-4-piperidinyl)-1H-benzimidazole, 1H-Benzimidazole','2-(1-nitroso-4-piperidinyl)- (ACI), 2-(1-Nitrosopiperidin-4-yl)-1H-1,3-benzodiazole, 2-(1-Nitroso-4-piperidinyl)-1H-benzimidazole |
| InChI Formula | InChI=1S/C12H14N4O/c17-15-16-7-5-9(6-8-16)12-13-10-3-1-2-4-11(10)14-12/h1-4,9H,5-8H2,(H,13,14) |
| SMILES | O=NN1CCC(CC1)c2nc3ccccc3[nH]2 |
| IUPAC Name | 2-(1-nitrosopiperidin-4-yl)-1H-benzimidazole |
| Molecular Weight (g/mol) | 230.266 |
| Formula Weight | 230.117 |