Learn More
(1S,3R,4R)-Ethyl 3-((tert-Butoxycarbonyl)amino)-4-(2-((5-chloropyridin-2-yl)amino)-2-oxoacetamido)cyclohexanecarboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | (1S,3R,4R)-Ethyl 3-((tert-Butoxycarbonyl)amino)-4-(2-((5-chloropyridin-2-yl)amino)-2-oxoacetamido)cyclohexanecarboxylate |
| CAS | 2810299-43-3 |
| Molecular Formula | C21H29ClN4O6 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C21H29ClN4O6/c1-5-31-19(29)12-6-8-14(15(10-12)25-20(30)32-21(2,3)4)24-17(27)18(28)26-16-9-7-13(22)11-23-16/h7,9,11-12,14-15H,5-6,8,10H2,1-4H3,(H,24,27)(H,25,30)(H,23,26,28)/t12-,14+,15+/m0/s1 |
| SMILES | O=C(OCC)[C@H]1CC[C@@H](NC(C(NC2=NC=C(Cl)C=C2)=O)=O)[C@H](NC(OC(C)(C)C)=O)C1 |
| IUPAC Name | ethyl (1S,3R,4R)-3-((tert-butoxycarbonyl)amino)-4-(2-((5-chloropyridin-2-yl)amino)-2-oxoacetamido)cyclohexane-1-carboxylate |
| Molecular Weight (g/mol) | 468.93 |
| Formula Weight | 468.18 |