Learn More
[(1S)-2-[[(tert-Butyl)dimethylsilyl]oxy]-1-(hydroxymethyl)ethyl]-carbamic Acid tert-Butyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | ((1S)-2-(((tert-Butyl)dimethylsilyl)oxy)-1-(hydroxymethyl)ethyl)-carbamic Acid tert-Butyl Ester |
| CAS | 185692-85-7 |
| Molecular Formula | C14H31NO4Si |
| Quantity | 5 g |
| Synonym | [(1S)-2-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-1-(hydroxymethyl)ethyl]-carbamic Acid 1,1-Dimethylethyl Ester, (S)-2-(tert-Butoxycarbonylamino)-3-[(tert-butyldimethylsilyl)oxy]propan-1-ol, (S)-tert-Butyl [1-(tert-butyldimethylsilyloxy)-3-hydroxypropan-2-yl]carbamate |
| InChI Formula | InChI=1S/C14H31NO4Si/c1-13(2,3)19-12(17)15-11(9-16)10-18-20(7,8)14(4,5)6/h11,16H,9-10H2,1-8H3,(H,15,17)/t11-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)N[C@@H](CO)CO[Si](C)(C)C(C)(C)C |
| IUPAC Name | tert-butyl N-[(2S)-1-[tert-butyl(dimethyl)silyl]oxy-3-hydroxypropan-2-yl]carbamate |
| Molecular Weight (g/mol) | 305.49 |
| Formula Weight | 305.2022 |