missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BS 5609 Marine Labels for BradyJet™ J7300 Color Label Printer

Product Code. 30058016
Click to view available options
Size:
104.90 mm (W) x 154.94 mm (H)
55.12 mm (W) x 85.09 mm (H)
57.15 mm (W) x 45.72 m (L)
74.93 mm (W) x 109.98 mm (H)
Unit Size:
1 roll
This item is not returnable. View return policy

Product Code. 30058016

Brand: Brady™ 170470

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Labels are BS 5609 Sections 2 and 3 certified to ensure identification stays compliant, stuck and seen during marine transport.

  • Ideal for GHS, hazardous materials, drum labelling, marine, transportation (DOT), storage and pharmaceutical labelling
  • Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup and seamless software interaction
  • For use with the BradyJet™ J7300 Color Label Printer
  • Ink cartridges are required to print on this material, use part numbers P-CMY-1 and P-K-1

Specifications

Certifications/Compliance BS 5609:1986 Section 2 and section 3, Compliant with Directive 2011/65/EU and 2015/863
Description BS 5609 Marine Labels for BradyJet J7300 Color Label Printer
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Polypropylene
Printable Sides Front
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard communication identification, Marine labelling, Safety identification
For Use With (Equipment) J7300
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 300 Labels/Box
Quantity 1
Shape Rectangle
Size 74.93 mm (W) x 109.98 mm (H)
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0088 in.
Thickness (Metric) 0.224 mm
Dimensions (LxW) 109.98 x 74.93 mm
Length (English) 4.33 in.
Length (Metric) 109.98 mm
Width (English) 2.95 in.
Width (Metric) 74.93 mm
Width (English) Printable Area 74.93 mm
Height (English) 4.33 in.
Height (Metric) 109.98 mm
Alternate Part Number J-261-2585
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.