missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(-)-Menthyl glyoxylate monohydrate, 98%

Product Code. 11364076
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364076 5 g 5g
11374076 25 g 25g
11384076 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11364076 Supplier Thermo Scientific Alfa Aesar Supplier No. L19208.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111969-64-3 | C12H22O4 | 230.304 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111969-64-3
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00792503
InChI Key BWZMJRSMHQDFIT-KXUCPTDWSA-N
Synonym 1r,2s,5r-2-isopropyl-5-methylcyclohexyl 2,2-dihydroxyacetate, l-menthyl glyoxylate hydrate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl dihydroxy-acetate, l-menthyl glyoxylate, l-menthylglyoxylate hydrate, --menthyl 2,2-dihydroxyacetate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl di, 2,2-dihydroxyacetic acid 1r,3r,4s-p-menth-3-yl ester, 1r,2s,5r-5-methyl-2-propan-2-ylcyclohexyl 2,2-dihydroxyacetate, 2,2-dihydroxyacetic acid 1r,2s,5r-2-isopropyl-5-methylcyclohexyl ester
PubChem CID 7373179
IUPAC Name [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2,2-dihydroxyacetate
SMILES CC1CCC(C(C1)OC(=O)C(O)O)C(C)C

Specifications

Melting Point 76°C to 80°C
Density 1.182
Boiling Point 95°C to 96°C (0.3 mmHg)
Quantity 5 g
UN Number UN3077
Beilstein 5637371
Sensitivity Air Sensitive
Optical Rotation −75° (c=1 in Methanol)
Formula Weight 230.31 (212.29 Anhydrous)
Percent Purity 98%
Chemical Name or Material (1R)-(-)-Menthyl glyoxylate monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.