missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (1R)-(-)-Camphorquinone, 98%

Product Code. p-7027384
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11451097

Brand: Alfa Aesar™ B24092.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10334-26-6
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00064160,MFCD00082863,MFCD00064160,MFCD00082863
InChI Key VNQXSTWCDUXYEZ-LDWIPMOCSA-N
Synonym 1r,4s-1,7,7-trimethylnorbornane-2,3-dione, 1r,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, 1s,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione
PubChem CID 16211789
IUPAC Name (4R)-4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
SMILES CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2=O

Specifications

Melting Point 197°C to 202°C
Quantity 1 g
Beilstein 2327696
Optical Rotation −102° (c=2 in Ethanol)
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material (1R)-(-)-Camphorquinone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.