Learn More
(1R,2S,5R)-(-)-Menthol-d4, TRC
Specifications
Specifications
| Chemical Name or Material | (1R,2S,5R)-(-)-Menthol-d4 |
| CAS | 1450828-98-4 |
| Molecular Formula | C10H16D4O |
| Quantity | 5 mg |
| Synonym | (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexanol-d4, [1R-(1α,2β,5α)]-5-Methyl-2- |
| InChI Formula | InChI=1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1/i6D2,9D,10D |
| SMILES | O[C@@]1([2H])[C@@](C(C)C)([2H])CC[C@@H](C)C1([2H])[2H] |
| IUPAC Name | (1R,2S,5R)-2-isopropyl-5-methylcyclohexan-1,2,6,6-d4-1-ol |
| Molecular Weight (g/mol) | 160.29 |
| Formula Weight | 160.18 |