missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-N-Methylsulfonyl-1,2-diphenylethanediamine, 98+%, Thermo Scientific Chemicals

Product Code. 11382809
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11382809

Brand: Thermo Scientific Alfa Aesar H27704.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 511534-44-4
Molecular Formula C15H18N2O2S
Molecular Weight (g/mol) 290.381
MDL Number MFCD03791036
InChI Key FSRRNSLQEDUDTP-HUUCEWRRSA-N
Synonym (1R,2R)-N-Methanesulfonyl-1,2-diphenylethanediamine
PubChem CID 11353383
IUPAC Name N-[(1R,2R)-2-amino-1,2-diphenylethyl]methanesulfonamide
SMILES CS(=O)(=O)NC(C1=CC=CC=C1)C(C2=CC=CC=C2)N

Specifications

Melting Point 117°C to 119°C
Quantity 1 g
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 290.38
Percent Purity ≥98%
Chemical Name or Material (1R,2R)-N-Methylsulfonyl-1,2-diphenylethanediamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.