missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, AISI 420FS, S42020, 100g, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Stainless Steel, AISI 420FS, S42020, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Titanium/Aluminium/Tin/Arsenic/Oxygen/Tungsten/Niobium/Cobalt/Phosphorus/Vanadium/Nitrogen/Copper/Nickel/Carbon/Silicon/Molybdenum/Manganese/Chromium/Lead/Zirconium/Calcium/Cerium/Magnesium/Zinc/Tantalum/Antimony/Selenium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0007 %/Titanium 0.0015 %/Aluminium 0.0025 %/Tin 0.0046 %/Arsenic 0.005 %/Oxygen 0.005 %/Tungsten 0.005 %/Niobium 0.012 %/Cobalt 0.016 %/Phosphorus 0.0182 %/Vanadium 0.028 %/Nitrogen 0.029 %/Copper 0.148 %/Nickel 0.269 %/Carbon 0.341 %/Silicon 0.357 %/Molybdenum 0.38 %/Manganese 1.13 %/Chromium 13.11 %/Lead (<0.001) %/Zirconium (<0.003) %/Calcium (<0.005) %/Cerium (<0.005) %/Magnesium (<0.009) %/Zinc (<0.02) %/Tantalum (0.0007) %/Antimony (0.004) %/Selenium (0.006) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction