missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Si/Cu, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Aluminum, Al/Si/Cu, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Tin/Magnesium/Lead/Zinc/Copper/Iron/Titanium/Nickel/Chromium/Manganese/Silicon |
| CAS | 7429-90-5 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Tin 0.006 %/Magnesium 0.009 %/Lead 0.014 %/Zinc 0.023 %/Copper 0.024 %/Iron 0.17 %/Titanium 0.33 %/Nickel 0.41 %/Chromium 0.46 %/Manganese 1.16 %/Silicon 1.65 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction