missing translation for 'onlineSavingsMsg'
Learn More

Chloroquine-d4 Phosphate Salt, TRC

Product Code. 30361030
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 30361030

Brand: Toronto Research Chemicals TRCC3799671MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloroquine-d4 Phosphate Salt
Purity Grade Notes HPLC
Molecular Formula C18H22D4ClN3 •xH3PO4
Quantity 1 mg
Synonym N4-(7-Chloro-4-quinolinyl)-N1,N1-diethyl-1,4-pentanediamine-d4 Phosphate Salt, Aralen-d4 Phosphate Salt, Aralen-d4 Phosphate, Arechin-d4, Avloclor-d4, Bemaphate-d4, Chingamin-d4, Chingamin-d4 Phosphate, SN 7618-d4, Sanoquin-d4, Tanakan-d4
InChI Formula InChI=1S/C18H26ClN3/c1-4-22(5-2)12-6-7-14(3)21-17-10-11-20-18-13-15(19)8-9-16(17)18/h8-11,13-14H,4-7,12H2,1-3H3,(H,20,21)/i6D2,12D2
SMILES ClC1=CC=C2C(N=CC=C2NC(CC([2H])(C([2H])(N(CC)CC)[2H])[2H])C)=C1
IUPAC Name 4-N-(7-chloroquinolin-4-yl)-1,1,2,2-tetradeuterio-1-N,1-N-diethylpentane-1,4-diamine
Molecular Weight (g/mol) 323.9098
Percent Purity >95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.