missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-1-(N-methylcarboxamidine) hydrochloride, 96%, Thermo Scientific™

Product Code. 15437775
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437775 25 g 25g
15427775 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437775 Supplier Thermo Scientific Alfa Aesar Supplier No. H30607.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194852-88-5
Molecular Formula C5H9ClN4
Molecular Weight (g/mol) 160.605
MDL Number MFCD11114408
InChI Key SYSWHVMZAHBYJO-UHFFFAOYSA-N
Synonym 1h-pyrazole-1-n-methylcarboxamidine hydrochloride, n'-methylpyrazole-1-carboximidamide hydrochloride, e-n'-methylpyrazole-1-carboximidamide hydrochloride, n1-methyl-1h-pyrazole-1-carboxamidine hydrochloride
PubChem CID 73995114
IUPAC Name N'-methylpyrazole-1-carboximidamide;hydrochloride
SMILES CN=C(N)N1C=CC=N1.Cl

Specifications

Melting Point 156°C to 159°C
Quantity 25 g
Sensitivity Hygroscopic
Formula Weight 160.61
Percent Purity 96%
Chemical Name or Material 1H-Pyrazole-1-(N-methylcarboxamidine) hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.