missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazole, 99%

Product Code. p-7022579
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498406 50 g 50g
11408416 250 g 250g
11488406 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498406 Supplier Thermo Scientific Alfa Aesar Supplier No. A15423.18
Add to basket
This item is not returnable. View return policy

CAS: 95-14-7 | C6H5N3 | 119.127 g/mol

1H-Benzotriazole is an anticorrosive agent, which is useful in aircraft deicing and antifreeze fluids. It is also employed in dishwasher detergents. Further, it is used as a restrainer in photographic emulsions and also useful as a reagent for the determination of silver in analytical chemistry. It also serves as a corrosion inhibitor in the atmosphere and underwater. Further, it is utilized in the synthesis of amines from glyoxal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Benzotriazole is an anticorrosive agent, which is useful in aircraft deicing and antifreeze fluids. It is also employed in dishwasher detergents. Further, it is used as a restrainer in photographic emulsions and also useful as a reagent for the determination of silver in analytical chemistry. It also serves as a corrosion inhibitor in the atmosphere and underwater. Further, it is utilized in the synthesis of amines from glyoxal.

Solubility
Soluble in water, benzene, toluene, chloroform, ethanol and N,N-dimethylformamide.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and heavy metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-14-7
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD00005699
InChI Key QRUDEWIWKLJBPS-UHFFFAOYSA-N
Synonym 1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
PubChem CID 7220
ChEBI CHEBI:75331
IUPAC Name 2H-benzotriazole
SMILES C1=CC2=NNN=C2C=C1

Specifications

Melting Point 96°C to 99°C
Density 1.360
Boiling Point 204°C (15 mm)
Flash Point 212°C (413°F)
Assay Percent Range 99%
Quantity 50 g
Beilstein 112133
Merck Index 14,1108
Solubility Information Soluble in water,benzene,toluene,chloroform,ethanol and N,N-dimethylformamide.
Formula Weight 119.13
Percent Purity 99%
Chemical Name or Material Benzotriazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1H-Benzotriazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 1H-Benzotriazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.