Learn More
L-Alanine-15N-N-t-BOC, CDN
Product Code. 30328140
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328140 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30328140 Supplier CDN Supplier No. CDNN38011G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-alanine-15 N
CAS 139952-87-7
Molecular Formula C8 H15 15 N O4
Quantity 1 g
Synonym L-Alanine-15 N, N-[(1,1-dimethylethoxy)carbonyl]- (9 CI, ACI), N-[(1,1-Dimethylethoxy)carbonyl]-L-alanine-15 N (ACI), -2-((tert-Butoxycarbonyl)amino-15 N)propanoic acid, (2 S)-2-(tert-Butoxycarbonylamino-15 N)propionic acid, (2 S)-2-[(2-Methylpropan-2-yl)oxycarbonylamino-15 N]propanoic acid, N-tert-Boc-L-alanine-15 N, N-tert-Boc-L-alanine-15 N, N-tert-Boc-L-alanine-15 N, N-tert-Boc-L-alanine-15 N
InChI Formula InChI=1 S/C8H15NO4/c1-5(6(10)11)9-7(12)13-8(2,3)4/h5H,1-4H3,(H,9,12)(H,10,11)/t5-/m0/s1/i9+1
SMILES C[C@H]([15 NH]C(=O)OC(C)(C)C)C(=O)O
IUPAC Name (2 S)-2-[(2-methylpropan-2-yl)oxycarbonyl(15 N)amino]propanoic acid
Molecular Weight (g/mol) 190.2024
Formula Weight 190.0971 g/mol
Percent Purity 99 atom % 15 N, min 98% Chemical Purity