Learn More
IRIS Biotech 1GR Fmoc-L-Hyp(tBu)-Wang Resin 1% DVB
Brand:  IRIS Biotech WAA5167.0001
Encompass
1GR Fmoc-L-Hyp(tBu)-Wang Resin 1% DVB
Product Code. 17289982
  • POA / 1g
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »