Learn More
Abnova™ 19p13/19q13 FISH Probe
Product Code. 16023335
Change view
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16023335 100 μL 100µL
16033335 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16023335 Supplier Abnova™ Supplier No. FG0207.100uL
Please to purchase this item. Need a web account? Register with us today!
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Specifications
Content And Storage Store at 4°C in the dark
Format Tube
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Product Type DNA/RNA
Quantity 100 μL
Form Liquid