Learn More
Bochem™ 18/10 Steel Dressing jars with lid
Specifications
Specifications
| Capacity (Metric) | 700 mL |
| Includes | Lid |
| Material | Stainless Steel |
| Quantity | 1Each |
| Height (Metric) | 100 mm |
| Closure Material | Stainless Steel |
| Diameter (Metric) | 100 mm |
| Closure Type | Lid |
| Shape | Round |
| Product Type | Metal Dressing Jar |