Learn More
Bochem™ 18/10 Stainless Steel Propeller Stirrers with 3 Fix Blades
Specifications
Specifications
| Diameter (Metric) | 70 mm |
| Diameter (Metric) Shaft | 8 mm |
| For Use With (Application) | Ideal for homogenizing and suspending |
| Includes | 3 Fix blades |
| Length (Metric) | 300 mm |
| Quantity | 1 Each |
| Type | Propeller Stirrer |