Learn More
17alpha-Boldenone, TRC
Specifications
Specifications
| Chemical Name or Material | 17alpha-Boldenone |
| CAS | 27833-18-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H26 O2 |
| Quantity | 100 mg |
| Synonym | Androsta-1,4-dien-3-one, 17-hydroxy-, (17α)- (9CI, ACI), (17α)-17-Hydroxyandrosta-1,4-dien-3-one (ACI), Androsta-1,4-dien-3-one, 17α-hydroxy- (8CI), 1,4-Androstadien-17α-ol-3-one, 17α-Boldenone, 17α-Hydroxyandrosta-1,4-dien-3-one, Epiboldenone, α-Boldenone |
| InChI Formula | InChI=1S/C19H26O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h7,9,11,14-17,21H,3-6,8,10H2,1-2H3/t14-,15-,16-,17+,18-,19-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@H]2O |
| IUPAC Name | (8R,9S,10R,13S,14S,17R)-17-hydroxy-10,13-dimethyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 286.4 |
| Show More |