Learn More
17-N-(2-Dimethylaminoethylamino)-17-demethoxy Geldanamycin Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 17-N-(2-Dimethylaminoethylamino)-17-demethoxy Geldanamycin Hydrochloride |
| CAS | 467214-21-7 |
| Molecular Formula | C32H49ClN4O8 |
| Quantity | 25 mg |
| Synonym | 17-Demethoxy-17-[[2-(dimethylamino)ethyl]amino]geldanamycin Hydrochloride, 17-N-(2-Dimethylaminoethylamino)-17-demethoxygeldanamycin Hydrochloride, 17-[2-(Dimethylamino)ethylamino]-17-desmethylgeldanamycin Hydrochloride, Alvespimycin Hydrochloride, DMAG Hydrochloride, NSC 707545 Hydrochloride, 17-DMAG Hydrochloride |
| InChI Formula | InChI=1S/C32H48N4O8.ClH/c1-18-14-22-27(34-12-13-36(5)6)24(37)17-23(29(22)39)35-31(40)19(2)10-9-11-25(42-7)30(44-32(33)41)21(4)16-20(3)28(38)26(15-18)43-8;/h9-11,16-18,20,25-26,28,30,34,38H,12-15H2,1-8H3,(H2,33,41)(H,35,40);1H/b11-9-,19-10+,21-16+;/t18-,20+,25+,26+,28-,30+;/m1./s1 |
| SMILES | Cl.CO[C@H]1C[C@H](C)CC2=C(NCCN(C)C)C(=O)C=C(NC(=O)\C(=C\C=C[C@H](OC)[C@@H](OC(=O)N)\C(=C\[C@H](C)[C@H]1O)\C)\C)C2=O |
| Molecular Weight (g/mol) | 653.21 |