Learn More
17-Methyl-5b-androstane-3a,17b-diol, TRC
Specifications
Specifications
| Chemical Name or Material | 17a-Methyl-5b-androstane-3a,17b-diol |
| CAS | 641-84-9 |
| Molecular Formula | C20H34O2 |
| Quantity | 500 mg |
| Synonym | Androstane-3,17-diol','17-methyl-','(3α,5β,17β)-, 5β-Androstane-3α,17β-diol','17-methyl- (7CI,8CI), (3α,5β,17β)-17-Methylandrostane-3,17-diol, 17-Methyl-5β-androstane-3α,17β-diol, 17α-Methyl-5β-androstan-3α,17β-diol, 17α-Methyl-5β-androstane-3α,17β-diol, AL 3841 |
| InChI Formula | InChI=1S/C20H34O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h13-17,21-22H,4-12H2,1-3H3/t13-,14-,15-,16+,17+,18+,19+,20+/m1/s1 |
| SMILES | C[C@]1(O)CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | (3R,5R,8R,9S,10S,13S,14S,17S)-10,13,17-trimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 306.4 |
| Formula Weight | 306.4828 |