Learn More
17-Iodo-androsta-5,16-diene-3Beta-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 17-Iodo-androsta-5,16-diene-3beta-ol |
| CAS | 32138-69-5 |
| Molecular Formula | C19H27IO |
| Quantity | 250 mg |
| Synonym | Androsta-5,16-dien-3-ol, 17-iodo-, (3β)-, Androsta-5,16-dien-3β-ol, 17-iodo- (7CI,8CI), (3β)-17-Iodoandrosta-5,16-dien-3-ol, 17-Iodoandrosta-5,16-diene-3β-ol, (3β)-17-Iodo-androsta-5,16-dien-3-ol |
| InChI Formula | InChI=1S/C19H27IO/c1-18-9-7-13(21)11-12(18)3-4-14-15-5-6-17(20)19(15,2)10-8-16(14)18/h3,6,13-16,21H,4-5,7-11H2,1-2H3/t13-,14-,15-,16-,18-,19-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CC=C4C[C@@H](O)CC[C@]34C)[C@@H]1CC=C2I |
| IUPAC Name | (3S,8R,9S,10R,13S,14S)-17-iodo-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 398.32 |
| Formula Weight | 398.1107 |